Tisk…

Zinc acetate, anhydrous ≥99.9%

Dodavatel: Thermo Fisher Scientific
Varování

H33734.09 H33734.18 H33734.30
H33734.09EA 1460 CZK
H33734.09 H33734.18 H33734.30
Zinc acetate, anhydrous ≥99.9%
Zinc acetate

Purity: 99.9+% CAS No.: 557-34-6 Molecular Formula: C4H6O4Zn Formula Weight: 183.47 Melting Point: 200° dec. UN No.: 3077 MDL No.: MFCD00012454

Vzorec: C₄H₆O₄Zn
MW: 183.48
Bod varu: 908
Bod tání: 200 °C (rozkl.)
Hustota: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012454
CAS číslo: 557-34-6
EINECS: 209-170-2
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR