Tisk…

D-(-)-Lactic acid

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid

J61871.MD J61871.03
J61871.MDEA 8480 CZK
J61871.MD J61871.03
D-(-)-Lactic acid
D-(-)-Lactic acid

CAS No.: 10326-41-7 Molecular Formula: C3H6O3 Formula Weight: 90.08 Storage Temperature: 2°C to 8°C Physical Form: Low melting solid Appearance: White to off-white Solubility: Soluble in water (50mg/ml) Beilstein Reference: 1720254 Merck Reference: 14,5335 Optical Rotation: -0.4° (c=8 in water) MDL No.: MFCD00068313


Useful chiral synthon, building block for depsipeptides

Vzorec: C₃H₆O₃
MW: 90,08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00068313
CAS číslo: 10326-41-7
EINECS: 233-713-2
Merck Index: 14,5335

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR