L(-)-Glutathione (oxidised form)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
CAS No.: 27025-41-8
Molecular Formula: C20H32N6O12S2
Formula Weight: 612.64
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 1718700
Merck Reference: 14,4475
Optical Rotation: -105° (c=2 in water)
MDL No.: MFCD00063106
Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH
Vzorec:
C₂₀H₃₂N₆O₁₂S₂ Bod tání: 185 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063106 CAS číslo: 27025-41-8 EINECS: 248-170-7 Merck Index: 14,4475 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...