Tisk…

Spironolaktón

Dodavatel: Thermo Fisher Scientific
Nebezpečí

J60119.03 J60119.06 J60119.09
J60119.03EA 1030 CZK
J60119.03 J60119.06 J60119.09
Spironolaktón
Spironolaktón

CAS No.: 52-01-7 Molecular Formula: C24H32O4S Formula Weight: 416.58 Physical Form: Powder Appearance: White Melting Point: 207-208° MDL No.: MFCD00082250


Antihypertensive potassium sparing diuretic; aldosterone receptor antagonist

Vzorec: C₂₄H₃₂O₄S
Bod varu: 597 °C (1013 hPa)
Bod tání: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00082250
CAS číslo: 52-01-7
EINECS: 200-133-6
Merck Index: 14,8760

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR