Tisk…

7-Hydroxychromone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 7-Hydroxy-4-chromone, 7-Hydroxy-4H-chromen-4-one

M-Clarity™ quality level = MQ100

683949-1G 683949-5G
SIAL683949-1GEA 1840 CZK
SIAL683949-1G SIAL683949-5G
7-Hydroxychromone, Sigma-Aldrich®
7-Hydroxychromone
Vzorec: C₉H₆O₃
MW: 162,1421 g/mol
MDL Number: MFCD00209371
CAS číslo: 59887-89-7
EINECS: 261-973-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR