Molybdenum boride ≥99% (metals basis)
Dodavatel: Thermo Scientific
12563.14
12563.22
12563.14EA
2780
CZK
12563.14
12563.22
Molybdenum boride ≥99% (metals basis)
Molybdenum boride
Purity: 99%
CAS No.: 12006-98-3
Molecular Formula: MoB
Formula Weight: 106.75
Physical Form: Powder
Vzorec:
BMo MW: 106,75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014219 CAS číslo: 12006-98-3 EINECS: 234-492-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...