Tisk…

Ditungsten pentaboride ≥99.4% (metals basis)

Dodavatel: Thermo Fisher Scientific

Synonyms: Tungsten boride

41664.14 41664.22
41664.14EA 2130 CZK
41664.14 41664.22
Ditungsten pentaboride ≥99.4% (metals basis)
Ditungsten pentaboride

Purity: 99.4% CAS No.: 12007-98-6 Molecular Formula: W2B5, may contain small amounts of other phases Formula Weight: 421.76 Physical Form: -325 Mesh Powder Odour: Odourless MDL No.: MFCD00049707

Vzorec: B₅W₂
MW: 421.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049707
CAS číslo: 12007-98-6
EINECS: 234-524-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR