Tisk…

N,N-Dimethylformamide dineopentyl acetal, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

140244-10G 140244-50G
SIAL140244-10GEA 0 CZK
SIAL140244-10G SIAL140244-50G
N,N-Dimethylformamide dineopentyl acetal, Sigma-Aldrich®
N,N-Dimethylformamide dineopentyl acetal
Vzorec: C₁₃H₂₉NO₂
MW: 231,38 g/mol
MDL Number: MFCD00008851
CAS číslo: 4909-78-8
EINECS: 225-536-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR