Tisk…

N,N-Dimethylformamide dineopentyl acetal ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L02728.09 L02728.18
L02728.09EA 1460 CZK
L02728.09 L02728.18
N,N-Dimethylformamide dineopentyl acetal ≥98%
N,N-Dimethylformamide dineopentyl acetal

Purity: 98% CAS No.: 4909-78-8 Molecular Formula: C13H29NO2 Formula Weight: 231.38 Flash Point: 63°(145°F) Refractive Index: 1.4117 Boiling Point: 85-87°/10mm Beilstein Reference: 741992 MDL No.: MFCD00008851

Vzorec: C₁₃H₂₉NO₂
MW: 231.38 g/mol
Bod varu: 85…87 °C (10 mmHg)
Hustota: 0.830
Bod vzplanutí: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008851
CAS číslo: 4909-78-8
EINECS: 225-536-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR