Tisk…

α,2-Dibromo-5-fluorotoluene ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L09553.03 L09553.06
L09553.03EA 2200 CZK
L09553.03 L09553.06
α,2-Dibromo-5-fluorotoluene ≥97%
α,2-Dibromo-5-fluorotoluene

Purity: 97% CAS No.: 112399-50-5 Molecular Formula: C7H5Br2F Formula Weight: 267.92 Refractive Index: 1.5900 Melting Point: 33-36° Boiling Point: 89-90°/1mm Beilstein Reference: 4177658 UN No.: 3265 MDL No.: MFCD00042187

Vzorec: C₇H₅Br₂F
Bod varu: 89…90 °C (1 mmHg)
Bod tání: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00042187
CAS číslo: 112399-50-5
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR