2-Fluoro-N-methylbenzylamine 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine
Purity: 95%
CAS No.: 399-30-4
Molecular Formula: C8H10FN
Formula Weight: 139.17
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.047
Refractive Index: 1.047
Sensitivity: Air Sensitive
Appearance: Colourless to tan
Boiling Point: 73°
MDL No.: MFCD04629633
Vzorec:
C₈H₁₀FN MW: 139,17 g/mol Bod varu: 73 °C Hustota: 1,047 Bod vzplanutí: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04629633 CAS číslo: 399-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...