Tisk…

Phenyl formate 95%

Dodavatel: Thermo Scientific
Varování

L00289.06 L00289.14
L00289.06EA 1950 CZK
L00289.06 L00289.14
Phenyl formate 95%
Phenyl formate

Purity: 95% CAS No.: 1864-94-4 Molecular Formula: HCOOC6H5 Formula Weight: 122.12 Flash Point: 71°(159°F) Refractive Index: 1.5120 Boiling Point: 72-74°/13mm Beilstein Reference: 507113 MDL No.: MFCD00014126

Vzorec: C₇H₆O₂
MW: 122,12 g/mol
Bod varu: 72…74 °C (13 mmHg)
Hustota: 1,090
Bod vzplanutí: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014126
CAS číslo: 1864-94-4
EINECS: 217-471-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR