Tisk…

Sodium hexanoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-caproicacidsodiumsalt, Sodium Hexanoate

M-Clarity™ quality level = MQ200

C4026-10G C4026-25G
SIALC4026-10GEA 4550 CZK
SIALC4026-10G SIALC4026-25G
Sodium hexanoate, Sigma-Aldrich®
Sodium hexanoate
Vzorec: C₆H₁₁NaO₂
MDL Number: MFCD00059056
CAS číslo: 10051-44-2
EINECS: 233-179-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR