3',5'-Dichloro-2'-hydroxyacetophenone ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L10006.06
L10006.14
L10006.06EA
1520
CZK
L10006.06
L10006.14
3',5'-Dichloro-2'-hydroxyacetophenone ≥99%
3',5'-Dichloro-2'-hydroxyacetophenone
Purity: 99%
CAS No.: 3321-92-4
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Melting Point: 95-98°
Boiling Point: 132-134°/18mm
MDL No.: MFCD00016417
Vzorec:
C₈H₆Cl₂O₂ Bod varu: 132…134 °C (18 mmHg) Bod tání: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016417 CAS číslo: 3321-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...