Tisk…

Tetrahydro-4H-thiopyran-4-one, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetrahydrothiopyran-4-one

M-Clarity™ quality level = MQ200

155160-1G 155160-5G
SIAL155160-1GEA 1630 CZK
SIAL155160-1G SIAL155160-5G
Tetrahydro-4H-thiopyran-4-one, Sigma-Aldrich®
Tetrahydro-4H-thiopyran-4-one
Vzorec: C₅H₈OS
MW: 116,18 g/mol
MDL Number: MFCD00006660
CAS číslo: 1072-72-6
EINECS: 214-015-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR