Tetrahydro-4H-thiopyran-4-one ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tetrahydrothiopyran-4-one
A15872.14
A15872.03
A15872.06
A15872.14EA
16510
CZK
A15872.14
A15872.03
A15872.06
Tetrahydro-4H-thiopyran-4-one ≥99%
Tetrahydro-4H-thiopyran-4-one
Purity: 99%
CAS No.: 1072-72-6
Molecular Formula: C5H8OS
Formula Weight: 116.18
Odour: Weak
Melting Point: 60-64°
MDL No.: MFCD00006660
Vzorec:
C₅H₈OS MW: 116,18 g/mol Bod tání: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006660 CAS číslo: 1072-72-6 EINECS: 214-015-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...