Tisk…
Gadolinium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Dodavatel: Thermo Fisher Scientific
Nebezpečí
40571.NAEA 1240 CZK
40571.NA 40571.NC 40571.NE
Gadolinium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Gadolinium(III) perchlorate
CAS No.: 14017-52-8 Molecular Formula: Gd(ClO4)3.6H2O Formula Weight: 563.69 (455.60anhy) Physical Form: Liquid Odour: Odourless UN No.: 3211 MDL No.: MFCD00150239
Vzorec: Cl₃GdO₁₂
MW: 455.602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150239
CAS číslo: 14017-52-8
EINECS: 237-837-8
UN: 3211
ADR: 5.1 II