D(+)-Galactosamine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
M-Clarity™ quality level = MQ200
G1639-100MG
G1639-1G
G1639-5G
SIALG1639-100MGEA
1860
CZK
SIALG1639-100MG
SIALG1639-1G
SIALG1639-5G
D(+)-Galactosamine hydrochloride, Sigma-Aldrich®
D-(+)-Galactosamine hydrochloride
Vzorec:
C₆H₁₄ClNO₅ MW: 215.64 g/mol |
MDL Number:
MFCD00135830 CAS číslo: 1772-03-8 EINECS: 217-198-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...