Triphenylantimony dichloride ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H55719.06
H55719.14
H55719.06EA
2020
CZK
H55719.06
H55719.14
Triphenylantimony dichloride ≥99%
Triphenylantimony dichloride
Vzorec:
C₁₈H₁₅Cl₂Sb Bod tání: 143…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013598 CAS číslo: 594-31-0 EINECS: 209-834-1 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...