4'-Acetamido-2'-methylacetophenone ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
N-(4-Acetyl-3-methylphenyl)acetamide
H26933.14
H26933.06
H26933.14EA
3920
CZK
H26933.14
H26933.06
4'-Acetamido-2'-methylacetophenone ≥97%
4'-Acetamido-2'-methylacetophenone
Purity: 97%
CAS No.: 34956-31-5
Molecular Formula: C11H13NO2
Formula Weight: 191.23
Melting Point: 137-140°
MDL No.: MFCD02258876
Vzorec:
C₁₁H₁₃NO₂ MW: 191.23 g/mol Bod tání: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258876 CAS číslo: 34956-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...