4-(Methylthio)phenol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Methylmercapto)phenol
H26225.09
H26225.18
H26225.30
H26225.09EA
2270
CZK
H26225.09
H26225.18
H26225.30
4-(Methylthio)phenol ≥98%
4-(Methylthio)phenol
Purity: 98%
CAS No.: 1073-72-9
Molecular Formula: C7H8OS
Formula Weight: 140.21
Odour: Stench
Melting Point: 85-87°
Boiling Point: 153-156°/20mm
MDL No.: MFCD00002351
Vzorec:
C₇H₈OS MW: 140,21 g/mol Bod varu: 153…156 °C (20 mmHg) Bod tání: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002351 CAS číslo: 1073-72-9 EINECS: 214-031-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...