Tisk…

4-Fluoro-2-methoxybenzylamine 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (4-Fluoro-2-methoxyphenyl)methanamine, (4-Fluoro- 2-methoxy-phenyl) methanamine

H26796.03 H26796.06
H26796.03EA 2940 CZK
H26796.03 H26796.06
4-Fluoro-2-methoxybenzylamine 97%
4-Fluoro-2-methoxybenzylamine

Purity: 97% CAS No.: 870563-60-3 Molecular Formula: C8H10FNO Formula Weight: 155.17 Refractive Index: 1.5240

Vzorec: C₈H₁₀FNO
MW: 155,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116361
CAS číslo: 870563-60-3
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR