2-Chloroethyl phenyl sulphide, Sigma-Aldrich®
Dodavatel: Merck
417602-25ML
417602-5ML
SIAL417602-25MLEA
4280
CZK
SIAL417602-25ML
SIAL417602-5ML
2-Chloroethyl phenyl sulphide, Sigma-Aldrich®
2-Chloroethyl phenyl sulphide
Vzorec:
C₈H₉ClS MW: 172,68 g/mol |
MDL Number:
MFCD00013691 CAS číslo: 5535-49-9 EINECS: 226-891-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...