2-Chloroethyl phenyl sulphide ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B23265.06
B23265.14
B23265.06EA
930
CZK
B23265.06
B23265.14
2-Chloroethyl phenyl sulphide ≥98%
2-Chloroethyl phenyl sulphide
Purity: 98%
CAS No.: 5535-49-9
Molecular Formula: C8H9ClS
Formula Weight: 172.68
Flash Point: 104°(220°F)
Refractive Index: 1.5830
Boiling Point: 245°
Beilstein Reference: 1907116
UN No.: 2922
MDL No.: MFCD00013691
Vzorec:
C₈H₉ClS MW: 172,68 g/mol Bod varu: 245 °C Hustota: 1,178 Bod vzplanutí: 104 °C (220 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013691 CAS číslo: 5535-49-9 EINECS: 226-891-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...