3,5-Dibromo-p-toluidine ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3,5-Dibromo-4-methylaniline
L13101.06
L13101.03
L13101.06EA
6520
CZK
L13101.06
L13101.03
3,5-Dibromo-p-toluidine ≥99%
3,5-Dibromo-p-toluidine
Purity: 99%
CAS No.: 13194-73-5
Molecular Formula: C7H7Br2N
Formula Weight: 264.96
Melting Point: 90-92°
Beilstein Reference: 2639285
Vzorec:
C₇H₇Br₂N MW: 264,96 g/mol Bod tání: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151806 CAS číslo: 13194-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...