Tisk…

4-Nonylphenylboronic acid

Dodavatel: Cayman Chemical

Synonyms: 4-N-Nonylphenylboronic acid, 4-n-Nonylbenzeneboronic acid, FAAH/MAGL-IN-4

13140-100 13140-250 13140-500 13140-1
CAYM13140-100EA 797 CZK
CAYM13140-100 CAYM13140-250 CAYM13140-500 CAYM13140-1
4-Nonylphenylboronic acid
4-Nonylphenylboronic acid
Vzorec: C₁₅H₂₅BO₂
MW: 248.18 g/mol
MDL Number: MFCD02093070
CAS číslo: 256383-45-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR