Tisk…

4-Butylphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

521493-1G 521493-5G
SIAL521493-1GEA 1680 CZK
SIAL521493-1G SIAL521493-5G
4-Butylphenylboronic acid, Sigma-Aldrich®
4-Butylphenylboronic acid
Vzorec: C₁₀H₁₅BO₂
MDL Number: MFCD02093926
CAS číslo: 145240-28-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR