Tisk…

4-tert-Butylphenylboronic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid

B24408.03 B24408.06
B24408.03EA 1270 CZK
B24408.03 B24408.06
4-tert-Butylphenylboronic acid ≥97%
4-tert-Butylphenylboronic acid

Purity: 97% CAS No.: 123324-71-0 Molecular Formula: C10H15BO2 Formula Weight: 178.04 Storage Temperature: 2°C to 8°C Melting Point: 191-196° Beilstein Reference: 5330959 MDL No.: MFCD01009697

Vzorec: C₁₀H₁₅BO₂
MW: 178,04 g/mol
Bod tání: 191…196 °C
MDL Number: MFCD01009697
CAS číslo: 123324-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR