Allyl methyl sulphide ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Allyl(methyl)sulphane
B22514.06
B22514.14
B22514.06EA
841
CZK
B22514.06
B22514.14
Allyl methyl sulphide ≥98%
Allyl methyl sulphide
Purity: 98%
CAS No.: 10152-76-8
Molecular Formula: CH2=CHCH2SCH3
Formula Weight: 88.17
Storage Temperature: 2°C to 8°C
Flash Point: 18°(65°F)
Refractive Index: 1.4714
Odour: Stench
Boiling Point: 91-93°
UN No.: 1993
MDL No.: MFCD00008657
Vzorec:
C₄H₈S MW: 88,17 g/mol Bod varu: 91…93 °C Hustota: 0,803 Bod vzplanutí: 18 °C (65 °F) |
MDL Number:
MFCD00008657 CAS číslo: 10152-76-8 EINECS: 233-422-0 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...