Tisk…

4'-(4-Bromophenyl)acetophenone ≥97%

Dodavatel: Thermo Scientific

Synonyms: 4-Acetyl-4'-bromobiphenyl, 1-(4'-Bromo-[1,1'-biphenyl]-4-yl)ethanone

L13798.14 L13798.22
L13798.14EA 2760 CZK
L13798.14 L13798.22
4'-(4-Bromophenyl)acetophenone ≥97%
4'-(4-Bromophenyl)acetophenone

Purity: 97% CAS No.: 5731-01-1 Molecular Formula: C14H11BrO Formula Weight: 275.15 Melting Point: 125-127° Beilstein Reference: 2048590

Vzorec: C₁₄H₁₁BrO
MW: 275,15 g/mol
Bod tání: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00143242
CAS číslo: 5731-01-1
EINECS: 227-236-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR