Tisk…

Zinc bromide hydrate ≥99.9% (metals basis)

Dodavatel: Thermo Scientific
Nebezpečí

44230.22 44230.36
44230.22EA 2380 CZK
44230.22 44230.36
Zinc bromide hydrate ≥99.9% (metals basis)
Zinc bromide hydrate

Purity: 99.9% Molecular Formula: ZnBr2.xH2O Formula Weight: 225.19(anhy) Physical Form: Crystalline Merck Reference: 14,10129 UN No.: 3260 MDL No.: MFCD00011294

Vzorec: Br₂Zn·H₂O
MW: 225,19 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS číslo: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 14,10129

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR