Tisk…

Zinc bromide hydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

12960.06 12960.22
12960.06EA 1770 CZK
12960.06 12960.22
Zinc bromide hydrate ≥99.999% (metals basis), Puratronic®
Zinc bromide hydrate

Purity: 99.999% CAS No.: 7699-45-8 Molecular Formula: ZnBr2.xH2O (x≈2) Formula Weight: 225.19(anhy) Physical Form: Crystalline UN No.: 3260 MDL No.: MFCD00011294

Vzorec: Br₂Zn·H₂O
MW: 225.19 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS číslo: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8 II
Merck Index: 14,10129

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR