Tisk…

Nickel chromite

Dodavatel: Thermo Scientific
Nebezpečí

19424.14 19424.22
19424.14EA 2900 CZK
19424.14 19424.22
Nickel chromite
Nickel chromite

Contains approximately 10% NiCrO₃. Dehydrogenation catalyst


CAS No.: 12018-18-7 Molecular Formula: Cr2NiO4 Formula Weight: 226.70 Physical Form: -100 Mesh Powder Odour: Odourless MDL No.: MFCD00064806

Vzorec: Cr₂NiO₄
MW: 226,70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064806
CAS číslo: 12018-18-7
EINECS: 234-636-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR