Tisk…

Niobium diboride ≥99% (metals basis)

Dodavatel: Thermo Fisher Scientific

Synonyms: Niobium boride

12573.14 12573.22
12573.14EA 2290 CZK
12573.14 12573.22
Niobium diboride ≥99% (metals basis)
Niobium diboride

Purity: 99% CAS No.: 12007-29-3 Molecular Formula: NbB2 Formula Weight: 114.53 Physical Form: Powder Odour: Odourless MDL No.: MFCD00016236

Vzorec: B₂Nb
MW: 114.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016236
CAS číslo: 12007-29-3
EINECS: 234-503-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR