Tisk…
Silicon nitride predominantly β-phase ≤10 µm, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Silicon(IV) nitride
M-Clarity™ quality level = MQ100
248622-100G 248622-500G
SIAL248622-100GEA 5010 CZK
SIAL248622-100G SIAL248622-500G
Silicon nitride predominantly β-phase ≤10 µm, Sigma-Aldrich®
Silicon nitride
Vzorec: N₄Si₃
MW: 140.28 g/mol
MDL Number: MFCD00011230
CAS číslo: 12033-89-5
EINECS: 234-796-8