Tisk…

Calcium nitride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

415103-100G 415103-25G
SIAL415103-100GEA 0 CZK
SIAL415103-100G SIAL415103-25G
Calcium nitride, Sigma-Aldrich®
Calcium nitride
Vzorec: Ca₃N₂
MW: 148,25 g/mol
MDL Number: MFCD00015985
CAS číslo: 12013-82-0
EINECS: 234-592-9
UN: 3178
ADR: 4.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR