Tisk…

2-Biphenylboronic acid pinacol ester ≥97%

Dodavatel: Thermo Fisher Scientific

H32320.03 H32320.06
H32320.03EA 1280 CZK
H32320.03 H32320.06
2-Biphenylboronic acid pinacol ester ≥97%
2-Biphenylboronic acid pinacol ester

Purity: 97% CAS No.: 914675-52-8 Molecular Formula: C18H21BO2 Formula Weight: 280.17 Melting Point: 76-79° MDL No.: MFCD14636459

Vzorec: C₁₈H₂₁BO₂
Bod tání: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD14636459
CAS číslo: 914675-52-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR