Tisk…

2,4,6-Trimethoxybenzylamine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

300985-5G
SIAL300985-5GEA 8720 CZK
SIAL300985-5G
2,4,6-Trimethoxybenzylamine hydrochloride, Sigma-Aldrich®
2,4,6-Trimethoxybenzylamine hydrochloride
Vzorec: C₁₀H₁₆ClNO₃
MDL Number: MFCD00012855
CAS číslo: 146548-59-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR