Tisk…

4-Nitrobenzylamine hydrochloride 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: (4-Nitrophenyl)methanamine hydrochloride

B21748.03 B21748.06 B21748.14
B21748.03EA 1460 CZK
B21748.03 B21748.06 B21748.14
4-Nitrobenzylamine hydrochloride 97%
4-Nitrobenzylamine hydrochloride

Purity: 97% CAS No.: 18600-42-5 Molecular Formula: C7H8N2O2.HCl Formula Weight: 188.62 Melting Point: 255-257° dec. Beilstein Reference: 3629994

Vzorec: C₇H₉ClN₂O₂
MW: 188,62 g/mol
Bod tání: 255…257 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012863
CAS číslo: 18600-42-5
EINECS: 242-441-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR