2-Bromo-4'-(trifluoromethoxy)acetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide
M-Clarity™ quality level = MQ100
669474-1G
669474-5G
SIAL669474-1GEA
2080
CZK
SIAL669474-1G
SIAL669474-5G
2-Bromo-4'-(trifluoromethoxy)acetophenone, Sigma-Aldrich®
2-Bromo-4'-(trifluoromethoxy)acetophenone
Vzorec:
C₉H₆BrF₃O₂ MW: 283.04 g/mol |
MDL Number:
MFCD00052333 CAS číslo: 103962-10-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...