Tisk…
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: 4-Aminomethylphenylboronic acid hydrochloride, 4-(Aminomethyl)benzeneboronic acid hydrochloride
H52855.03 H52855.06
H52855.03EA 5050 CZK
H52855.03 H52855.06
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
4-(Aminomethyl)phenylboronic acid hydrochloride
Purity: 96% CAS No.: 75705-21-4 Molecular Formula: C7H10BNO2.HCl Formula Weight: 187.43 Melting Point: 240-250° MDL No.: MFCD01632199
Vzorec: C₇H₁₁BClNO₂
MW: 187.43 g/mol
Bod tání: 240…250 °C
Storage Temperature: Ambient
MDL Number: MFCD01632199
CAS číslo: 75705-21-4