Tisk…

N,N-Bis(trifluoromethanesulphonyl)aniline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)

78175-25G 78175-50G 78175-5G
SIAL78175-25GEA 9980 CZK
SIAL78175-25G SIAL78175-50G SIAL78175-5G
N,N-Bis(trifluoromethanesulphonyl)aniline, Sigma-Aldrich®
N,N-Bis(trifluoromethanesulphonyl)aniline
Vzorec: C₈H₅F₆NO₄S₂
MW: 357,25 g/mol
MDL Number: MFCD00000404
CAS číslo: 37595-74-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR