Tisk…

3-Fluoro-(4-methoxycarbonyl)phenylboronic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: (3-Fluoro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 3-Fluoro-4-methoxycarbonylphenylboronic acid, 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

H52510.03 H52510.06
H52510.03EA 4260 CZK
H52510.03 H52510.06
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid ≥97%
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid

Purity: 97% CAS No.: 505083-04-5 Molecular Formula: C8H8BFO4 Formula Weight: 197.96 Melting Point: 180-184° MDL No.: MFCD04115665

Vzorec: C₈H₈BFO₄
MW: 197,96 g/mol
Bod tání: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD04115665
CAS číslo: 505083-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR