Tisk…

4-Amino-3-chlorophenol hydrochloride 98%

Dodavatel: Thermo Fisher Scientific
Varování

A19720.06 A19720.14 A19720.22 AB124976.0005
A19720.06EA 1320 CZK
A19720.06 A19720.14 A19720.22 ABCRAB124976.0005
4-Amino-3-chlorophenol hydrochloride 98%
4-Amino-3-chlorophenol hydrochloride

Purity: 98% CAS No.: 52671-64-4 Molecular Formula: C6H6ClNO.HCl Formula Weight: 180.04 Melting Point: 254-256° dec. MDL No.: MFCD00143110

Vzorec: C₆H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00143110
CAS číslo: 52671-64-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR