3-Carboxy-5-nitrophenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid
H29348.03
H29348.06
H29348.03EA
1450
CZK
H29348.03
H29348.06
3-Carboxy-5-nitrophenylboronic acid ≥97%
3-Carboxy-5-nitrophenylboronic acid
Purity: 97%
CAS No.: 101084-81-5
Molecular Formula: C7H6BNO6
Formula Weight: 210.94
MDL No.: MFCD00757433
Vzorec:
C₇H₆BNO₆ MW: 210,94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00757433 CAS číslo: 101084-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...