Tisk…

4-(Ethoxycarbonyl)phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

M-Clarity™ quality level = MQ100

574643-1G 574643-5G
SIAL574643-1GEA 2790 CZK
SIAL574643-1G SIAL574643-5G
4-(Ethoxycarbonyl)phenylboronic acid, Sigma-Aldrich®
4-(Ethoxycarbonyl)phenylboronic acid
Vzorec: C₉H₁₁BO₄
MW: 193,99 g/mol
MDL Number: MFCD02179441
CAS číslo: 4334-88-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR