2-(Ethoxycarbonyl)phenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid
H25867.06
H25867.03
H25867.06EA
3400
CZK
H25867.06
H25867.03
2-(Ethoxycarbonyl)phenylboronic acid ≥98%
2-(Ethoxycarbonyl)phenylboronic acid
Purity: 98%
CAS No.: 380430-53-5
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 130-133°
MDL No.: MFCD02179453
Vzorec:
C₉H₁₁BO₄ Bod tání: 130…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179453 CAS číslo: 380430-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...