Tisk…

(S)-(-)-N-Benzyl-α-methylbenzylamine, Sigma-Aldrich®

Dodavatel: Merck

427470-10ML 427470-50ML
SIAL427470-10MLEA 2320 CZK
SIAL427470-10ML SIAL427470-50ML
(S)-(-)-N-Benzyl-α-methylbenzylamine, Sigma-Aldrich®
(S)-(-)-N-Benzyl-α-methylbenzylamine
Vzorec: C₁₅H₁₇N
MDL Number: MFCD00066325
CAS číslo: 17480-69-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR