Tisk…

Methyl propyl disulphide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W320102-100G-K W320102-1KG-K W320102-25G-K
SIALW320102-100G-KEA 4850 CZK
SIALW320102-100G-K SIALW320102-1KG-K SIALW320102-25G-K
Methyl propyl disulphide, Sigma-Aldrich®
Methyl propyl disulphide
Vzorec: C₄H₁₀S₂
MDL Number: MFCD00013400
CAS číslo: 2179-60-4
EINECS: 218-551-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR