Tisk…

Allyl methyl carbonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Carbonic acid Allyl Methyl Ester, Allyl Methyl Carbonate

381381-25ML 381381-5ML
SIAL381381-25MLEA 11900 CZK
SIAL381381-25ML SIAL381381-5ML
Allyl methyl carbonate, Sigma-Aldrich®
Allyl methyl carbonate
Vzorec: C₅H₈O₃
MW: 116,1152 g/mol
MDL Number: MFCD00134545
CAS číslo: 35466-83-2
EINECS: 468-750-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR